Assay Details
| Gene Symbol: Tmem245 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175518.5 | NP_780727.3 | 1 - 2 | 580 | 63 | |
| GenBank mRNA | AK052826.1 | - | 1 - 2 | 575 | 63 | |
| BB810057.1 | - | 1 - 2 | 237 | 63 | ||
| CJ067096.1 | - | 1 - 2 | 581 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmem245
Genomic Map