Assay Details
| Gene Symbol: Dgkk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177914.3 | NP_808582.3 | 1283 | 157 | ||
| GenBank mRNA | BC075627.1 | - | 1283 | 6852516 | 157 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dgkk
Genomic Map