Assay Details
| Gene Symbol: Sh3rf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021506.2 | NP_067481.2 | 9 - 10 | 2066 | 80 | |
| GenBank mRNA | AF030131.1 | - | 9 - 10 | 1968 | 80 | |
| AK045470.1 | - | 7 - 8 | 1201 | 80 | ||
| AK148137.1 | - | 9 - 10 | 1997 | 80 | ||
| AK173185.1 | - | 9 - 10 | 2432 | 80 | ||
| BC060113.1 | - | 10 - 11 | 1938 | 6405370 | 80 | |
| BC060696.1 | - | 10 - 11 | 1938 | 6405370 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sh3rf1
Genomic Map