Assay Details
| Gene Symbol: Mef2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025282.3 | NP_079558.1 | 4 - 5 | 835 | 95 | |
| GenBank mRNA | AK009139.1 | - | 4 - 5 | 675 | 95 | |
| BC037731.1 | - | 3 - 4 | 563 | 4500786 | 95 | |
| BC057650.1 | - | 3 - 4 | 552 | 4500786 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mef2c
Genomic Map