Assay Details
| Gene Symbol: Mef2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170537.1 | NP_001164008.1 | 6 - 7 | 1161 | 62 | |
| NM_025282.3 | NP_079558.1 | 6 - 7 | 1155 | 62 | ||
| GenBank mRNA | AK009139.1 | - | 6 - 7 | 995 | 62 | |
| AK132573.1 | - | 6 - 7 | 990 | 62 | ||
| BC026841.1 | - | 5 - 6 | 893 | 4481227 | 62 | |
| BC037731.1 | - | 5 - 6 | 883 | 4500786 | 62 | |
| BC057650.1 | - | 5 - 6 | 872 | 4500786 | 62 | |
| L13171.1 | - | 4 - 5 | 722 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mef2c
Genomic Map