Assay Details
| Gene Symbol: Gla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013463.2 | NP_038491.2 | 5 - 6 | 880 | 99 | |
| GenBank mRNA | AK040081.1 | - | 5 - 6 | 812 | 99 | |
| AK054547.1 | - | 5 - 6 | 832 | 99 | ||
| AK145011.1 | - | 5 - 6 | 822 | 99 | ||
| AK145148.1 | - | 880 | 99 | |||
| AK164231.1 | - | 5 - 6 | 813 | 99 | ||
| AK166204.1 | - | 5 - 6 | 813 | 99 | ||
| AK166245.1 | - | 5 - 6 | 829 | 99 | ||
| BC009021.1 | - | 5 - 6 | 833 | 3708950 | 99 | |
| U34071.1 | - | 5 - 6 | 830 | 99 | ||
| U50716.1 | - | 5 - 6 | 818 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gla
Genomic Map