Assay Details
| Gene Symbol: Cenpi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK043344.1 | - | 10 - 11 | 1114 | 94 | |
| AK145200.1 | - | 11 - 12 | 94 | |||
| AK161473.1 | - | 11 - 12 | 1253 | 94 | ||
| AK166305.1 | - | 7 - 8 | 94 | |||
| BC030328.1 | - | 11 - 12 | 1170 | 5376020 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cenpi
Genomic Map