Assay Details
| Gene Symbol: AI314180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172381.2 | NP_759013.2 | 24 - 25 | 3008 | 68 | |
| GenBank mRNA | AK043020.1 | - | 25 - 26 | 2925 | 68 | |
| AK129127.1 | - | 7 - 8 | 948 | 68 | ||
| BC058684.1 | - | 24 - 25 | 3008 | 6834912 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

AI314180
Genomic Map