Assay Details
| Gene Symbol: AI314180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172381.2 | NP_759013.2 | 25 - 26 | 3152 | 92 | |
| GenBank mRNA | AK035657.1 | - | 4 - 5 | 511 | 92 | |
| AK043020.1 | - | 26 - 27 | 3069 | 92 | ||
| AK129127.1 | - | 8 - 9 | 1092 | 92 | ||
| BC058684.1 | - | 25 - 26 | 3152 | 6834912 | 92 | |
More Information
Set Membership: |
|

AI314180
Genomic Map