Assay Details
| Gene Symbol: AI314180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172381.2 | NP_759013.2 | 28 - 29 | 3393 | 80 | |
| GenBank mRNA | AK035657.1 | - | 7 - 8 | 752 | 80 | |
| AK043020.1 | - | 29 - 30 | 3310 | 80 | ||
| AK129127.1 | - | 11 - 12 | 1333 | 80 | ||
| BC058684.1 | - | 28 - 29 | 3393 | 6834912 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

AI314180
Genomic Map