Assay Details
| Gene Symbol: AI314180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172381.2 | NP_759013.2 | 35 - 36 | 4188 | 66 | |
| GenBank mRNA | AK043020.1 | - | 36 - 37 | 4105 | 66 | |
| AK129127.1 | - | 18 - 19 | 2128 | 66 | ||
| BC025035.1 | - | 5 - 6 | 460 | 66 | ||
| BC058684.1 | - | 35 - 36 | 4188 | 6834912 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC025035.1 transcripts.
Set Membership: |
|

AI314180
Genomic Map