Assay Details
| Gene Symbol: AI314180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172381.2 | NP_759013.2 | 36 - 37 | 4253 | 69 | |
| GenBank mRNA | AK043020.1 | - | 37 - 38 | 4170 | 69 | |
| AK129127.1 | - | 19 - 20 | 2193 | 69 | ||
| BC025035.1 | - | 6 - 7 | 525 | 69 | ||
| BC058684.1 | - | 36 - 37 | 4253 | 6834912 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

AI314180
Genomic Map