Assay Details
| Gene Symbol: AI314180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172381.2 | NP_759013.2 | 39 - 40 | 4501 | 112 | |
| GenBank mRNA | AK043020.1 | - | 40 - 41 | 4419 | 112 | |
| AK129127.1 | - | 22 - 23 | 2441 | 112 | ||
| BC025035.1 | - | 9 - 10 | 773 | 112 | ||
| BC058684.1 | - | 39 - 40 | 4501 | 6834912 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|

AI314180
Genomic Map