Assay Details
| Gene Symbol: AI314180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172381.2 | NP_759013.2 | 2 - 3 | 382 | 127 | |
| GenBank mRNA | AK038775.1 | - | 3 - 4 | 296 | 127 | |
| AK041579.1 | - | 2 - 3 | 408 | 127 | ||
| AK043020.1 | - | 3 - 4 | 299 | 127 | ||
| AK087690.1 | - | 3 - 4 | 466 | 127 | ||
| BC058684.1 | - | 2 - 3 | 382 | 6834912 | 127 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK087690.1 transcripts.
Set Membership: |
|

AI314180
Genomic Map