Assay Details
| Gene Symbol: AI314180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172381.2 | NP_759013.2 | 46 - 47 | 5455 | 69 | |
| GenBank mRNA | AK030369.1 | - | 6 - 7 | 884 | 69 | |
| AK129127.1 | - | 29 - 30 | 3395 | 69 | ||
| BC025035.1 | - | 16 - 17 | 1727 | 69 | ||
| BC058684.1 | - | 46 - 47 | 5455 | 6834912 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

AI314180
Genomic Map