Assay Details
| Gene Symbol: AI314180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172381.2 | NP_759013.2 | 3 - 4 | 493 | 69 | |
| GenBank mRNA | AK038775.1 | - | 4 - 5 | 407 | 69 | |
| AK041579.1 | - | 3 - 4 | 519 | 69 | ||
| AK043020.1 | - | 4 - 5 | 410 | 69 | ||
| AK087690.1 | - | 4 - 5 | 577 | 69 | ||
| BC058684.1 | - | 3 - 4 | 493 | 6834912 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK087690.1 transcripts.
Set Membership: |
|

AI314180
Genomic Map