Assay Details
| Gene Symbol: AI314180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172381.2 | NP_759013.2 | 5 - 6 | 760 | 74 | |
| GenBank mRNA | AK038775.1 | - | 6 - 7 | 674 | 74 | |
| AK041579.1 | - | 5 - 6 | 786 | 74 | ||
| AK043020.1 | - | 6 - 7 | 677 | 74 | ||
| BC058684.1 | - | 5 - 6 | 760 | 6834912 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

AI314180
Genomic Map