Assay Details
| Gene Symbol: AI314180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172381.2 | NP_759013.2 | 7 - 8 | 1076 | 59 | |
| GenBank mRNA | AK038775.1 | - | 8 - 9 | 990 | 59 | |
| AK041579.1 | - | 7 - 8 | 1102 | 59 | ||
| AK043020.1 | - | 8 - 9 | 993 | 59 | ||
| AK087690.1 | - | 8 - 9 | 1163 | 59 | ||
| BC058684.1 | - | 7 - 8 | 1076 | 6834912 | 59 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK087690.1 transcripts.
Set Membership: |
|

AI314180
Genomic Map