Assay Details
| Gene Symbol: Mctp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030174.2 | NP_084450.2 | 17 - 18 | 2034 | 98 | |
| GenBank mRNA | AK013379.1 | - | 4 - 5 | 640 | 98 | |
| AK047562.1 | - | 17 - 18 | 2035 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mctp1
Genomic Map