Assay Details
| Gene Symbol: Mctp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030174.2 | NP_084450.2 | 8 - 9 | 1088 | 81 | |
| GenBank mRNA | AK047562.1 | - | 8 - 9 | 1089 | 81 | |
| AK054478.1 | - | 4 - 5 | 461 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mctp1
Genomic Map