Assay Details
| Gene Symbol: Slc38a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172479.3 | NP_766067.2 | 3 - 4 | 265 | 68 | |
| GenBank mRNA | AK054485.1 | - | 3 - 4 | 252 | 68 | |
| AK077667.1 | - | 3 - 4 | 234 | 68 | ||
| AK155928.1 | - | 3 - 4 | 218 | 68 | ||
| AK158125.1 | - | 2 - 3 | 386 | 68 | ||
| BC152401.1 | - | 3 - 4 | 234 | 40057282 | 68 | |
| BY247469.1 | - | 3 - 4 | 255 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc38a5
Genomic Map