Assay Details
| Gene Symbol: Tbc1d25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166437.1 | NP_001159909.1 | 5 - 6 | 664 | 64 | |
| NM_172478.3 | NP_766066.2 | 4 - 5 | 871 | 64 | ||
| GenBank mRNA | AK040932.1 | - | 4 - 5 | 614 | 64 | |
| AK078145.1 | - | 4 - 5 | 872 | 64 | ||
| AK166256.1 | - | 4 - 5 | 610 | 64 | ||
| BC113777.1 | - | 5 - 6 | 612 | 64 | ||
| BC128565.1 | - | 5 - 6 | 614 | 64 | ||
| BC128566.1 | - | 4 - 5 | 821 | 40094900 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tbc1d25
Genomic Map