Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800017336.mm.1 |
NM_001081390.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017336.mm.1 |
NM_001081390.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017336.mm.1 |
TC0800002300.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800017337.mm.1 |
NM_001081390.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017337.mm.1 |
NM_001081390.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017337.mm.1 |
TC0800002300.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800017338.mm.1 |
NM_001081390.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017338.mm.1 |
NM_001081390.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017338.mm.1 |
TC0800002300.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800017335.mm.1 |
NM_001081390.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017335.mm.1 |
NM_001081390.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017335.mm.1 |
TC0800002300.mm.1 |
MTA TC |