Assay Details
| Gene Symbol: Rpsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011029.4 | NP_035159.3 | 2 - 3 | 225 | 78 | |
| GenBank mRNA | AF140348.1 | - | 2 - 3 | 212 | 78 | |
| AK010423.1 | - | 2 - 3 | 205 | 78 | ||
| AK010985.1 | - | 2 - 3 | 207 | 78 | ||
| AK011041.1 | - | 2 - 3 | 207 | 78 | ||
| AK011043.1 | - | 2 - 3 | 207 | 78 | ||
| AK082935.1 | - | 2 - 3 | 204 | 78 | ||
| AK088954.1 | - | 2 - 3 | 205 | 78 | ||
| AK134224.1 | - | 2 - 3 | 205 | 78 | ||
| AK135488.1 | - | 2 - 3 | 207 | 78 | ||
| AK145713.1 | - | 2 - 3 | 202 | 78 | ||
| AK151123.1 | - | 2 - 3 | 204 | 78 | ||
| AK160551.1 | - | 2 - 3 | 204 | 78 | ||
| AK160625.1 | - | 2 - 3 | 204 | 78 | ||
| AK165219.1 | - | 2 - 3 | 205 | 78 | ||
| AK166697.1 | - | 2 - 3 | 204 | 78 | ||
| AK166874.1 | - | 2 - 3 | 207 | 78 | ||
| AK167132.1 | - | 2 - 3 | 205 | 78 | ||
| AK205896.1 | - | 1 - 2 | 110 | 78 | ||
| AK211265.1 | - | 1 - 2 | 110 | 78 | ||
| AK212093.1 | - | 1 - 2 | 110 | 78 | ||
| AK212679.1 | - | 1 - 2 | 110 | 78 | ||
| AK213565.1 | - | 1 - 2 | 110 | 78 | ||
| AK214711.1 | - | 1 - 2 | 110 | 78 | ||
| AK215661.1 | - | 1 - 2 | 110 | 78 | ||
| AK216125.1 | - | 1 - 2 | 110 | 78 | ||
| AK218136.1 | - | 1 - 2 | 110 | 78 | ||
| AK219743.1 | - | 1 - 2 | 110 | 78 | ||
| AK219820.1 | - | 1 - 2 | 110 | 78 | ||
| AV586849.1 | - | 2 - 3 | 302 | 78 | ||
| BC003829.1 | - | 2 - 3 | 189 | 3600738 | 78 | |
| BC037195.1 | - | 1 - 2 | 152 | 4500748 | 78 | |
| BC055886.1 | - | 1 - 2 | 359 | 4240083 | 78 | |
| BC081461.1 | - | 1 - 2 | 160 | 6820702 | 78 | |
| BC084677.1 | - | 2 - 3 | 189 | 6476344 | 78 | |
| BC092041.1 | - | 2 - 3 | 190 | 2651126 | 78 | |
| BC094902.1 | - | 2 - 3 | 189 | 6433349 | 78 | |
| BC099601.1 | - | 2 - 3 | 190 | 1245698 | 78 | |
| BC110285.1 | - | 2 - 3 | 190 | 5151374 | 78 | |
| EU554633.1 | - | 1 - 2 | 127 | 78 | ||
| J02870.1 | - | 2 - 3 | 206 | 78 | ||
| X06406.1 | - | 2 - 3 | 204 | 78 | ||
More Information
Set Membership: |
|

Rpsa
Genomic Map