Assay Details
| Gene Symbol: Rpsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011029.4 | NP_035159.3 | 6 - 7 | 893 | 72 | |
| GenBank mRNA | AF140348.1 | - | 6 - 7 | 880 | 72 | |
| AK010423.1 | - | 6 - 7 | 873 | 72 | ||
| AK010985.1 | - | 6 - 7 | 875 | 72 | ||
| AK011041.1 | - | 6 - 7 | 875 | 72 | ||
| AK011043.1 | - | 6 - 7 | 875 | 72 | ||
| AK082935.1 | - | 6 - 7 | 872 | 72 | ||
| AK088954.1 | - | 6 - 7 | 873 | 72 | ||
| AK134224.1 | - | 6 - 7 | 873 | 72 | ||
| AK135488.1 | - | 6 - 7 | 875 | 72 | ||
| AK145713.1 | - | 6 - 7 | 943 | 72 | ||
| AK151123.1 | - | 6 - 7 | 872 | 72 | ||
| AK160551.1 | - | 6 - 7 | 872 | 72 | ||
| AK160625.1 | - | 6 - 7 | 872 | 72 | ||
| AK165219.1 | - | 6 - 7 | 873 | 72 | ||
| AK166697.1 | - | 6 - 7 | 872 | 72 | ||
| AK166874.1 | - | 6 - 7 | 875 | 72 | ||
| AK167132.1 | - | 6 - 7 | 873 | 72 | ||
| AK190545.1 | - | 1 - 2 | 101 | 72 | ||
| AK196804.1 | - | 1 - 2 | 100 | 72 | ||
| AK203733.1 | - | 1 - 2 | 101 | 72 | ||
| BC003829.1 | - | 6 - 7 | 857 | 3600738 | 72 | |
| BC037195.1 | - | 5 - 6 | 820 | 4500748 | 72 | |
| BC055886.1 | - | 5 - 6 | 1027 | 4240083 | 72 | |
| BC081461.1 | - | 5 - 6 | 828 | 6820702 | 72 | |
| BC084677.1 | - | 6 - 7 | 857 | 6476344 | 72 | |
| BC092041.1 | - | 6 - 7 | 858 | 2651126 | 72 | |
| BC094902.1 | - | 6 - 7 | 857 | 6433349 | 72 | |
| BC099601.1 | - | 6 - 7 | 858 | 1245698 | 72 | |
| BC110285.1 | - | 6 - 7 | 858 | 5151374 | 72 | |
| EU554633.1 | - | 5 - 6 | 795 | 72 | ||
| J02870.1 | - | 6 - 7 | 874 | 72 | ||
| X06406.1 | - | 6 - 7 | 872 | 72 | ||
More Information
Set Membership: |
|

Rpsa
Genomic Map