Assay Details
| Gene Symbol: Tgm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177911.4 | NP_808579.2 | 6 - 7 | 694 | 69 | |
| GenBank mRNA | AF486627.1 | - | 6 - 7 | 693 | 69 | |
| AK035279.1 | - | 6 - 7 | 695 | 69 | ||
| BC141297.1 | - | 6 - 7 | 691 | 9055909 | 69 | |
| BC145622.1 | - | 6 - 7 | 691 | 9054190 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tgm4
Genomic Map