Assay Details
| Gene Symbol: Tgm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177911.4 | NP_808579.2 | 9 - 10 | 1105 | 80 | |
| GenBank mRNA | AF486627.1 | - | 9 - 10 | 1104 | 80 | |
| AK035279.1 | - | 9 - 10 | 1106 | 80 | ||
| BC141297.1 | - | 9 - 10 | 1102 | 9055909 | 80 | |
| BC145622.1 | - | 9 - 10 | 1102 | 9054190 | 80 | |
More Information
Set Membership: |
|

Tgm4
Genomic Map