Assay Details
| Gene Symbol: Syn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110780.1 | NP_001104250.1 | 7 - 8 | 1108 | 65 | |
| NM_013680.4 | NP_038708.3 | 7 - 8 | 1108 | 65 | ||
| GenBank mRNA | BC022954.1 | - | 7 - 8 | 1086 | 5368098 | 65 |
| KJ174470.1 | - | 1 - 2 | 69 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Syn1
Genomic Map