Assay Details
| Gene Symbol: Slc6a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020049.4 | NP_064433.3 | 2 - 3 | 458 | 83 | |
| GenBank mRNA | AB033285.1 | - | 2 - 3 | 216 | 83 | |
| AF161714.1 | - | 2 - 3 | 458 | 83 | ||
| AF320226.1 | - | 2 - 3 | 402 | 83 | ||
| AK018553.1 | - | 2 - 3 | 317 | 83 | ||
| BC129908.1 | - | 2 - 3 | 249 | 40111716 | 83 | |
| BC129909.1 | - | 2 - 3 | 249 | 40111717 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc6a14
Genomic Map