Assay Details
| Gene Symbol: Araf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159645.1 | NP_001153117.1 | 1 - 2 | 78 | 71 | |
| NM_009703.2 | NP_033833.1 | 1 - 2 | 78 | 71 | ||
| GenBank mRNA | AK020547.1 | - | 1 - 2 | 44 | 71 | |
| AK039044.1 | - | 1 - 2 | 44 | 71 | ||
| AK085823.1 | - | 1 - 2 | 44 | 71 | ||
| AK135358.1 | - | 1 - 2 | 44 | 71 | ||
| AK163567.1 | - | 1 - 2 | 47 | 71 | ||
| AK169757.1 | - | 1 - 2 | 59 | 71 | ||
| BB857915.1 | - | 1 - 2 | 78 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK020547.1, AK039044.1, AK135358.1, AK163567.1, NM_001159645.1, NM_009703.2 transcripts.
Set Membership: |
|

Araf
Genomic Map