Assay Details
| Gene Symbol: Araf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009703.2 | NP_033833.1 | 7 - 8 | 830 | 62 | |
| GenBank mRNA | AK020547.1 | - | 7 - 8 | 796 | 62 | |
| AK034861.1 | - | 7 - 8 | 825 | 62 | ||
| AK085823.1 | - | 7 - 8 | 796 | 62 | ||
| AK135358.1 | - | 7 - 8 | 796 | 62 | ||
| AK163567.1 | - | 7 - 8 | 799 | 62 | ||
| AK169757.1 | - | 7 - 8 | 812 | 62 | ||
| BC004757.1 | - | 7 - 8 | 786 | 3154293 | 62 | |
| BC080721.1 | - | 1 - 2 | 268 | 62 | ||
| D00024.1 | - | 2 - 3 | 192 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Araf
Genomic Map