Assay Details
| Gene Symbol: Gm6938 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033482.1 | 1 - 2 | 172 | 58 | ||
| GenBank mRNA | BF470644.1 | - | 1 - 2 | 172 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gm6938
Genomic Map