Assay Details
| Gene Symbol: Stt3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024222.2 | NP_077184.2 | 4 - 5 | 1137 | 73 | |
| GenBank mRNA | AK012153.1 | - | 4 - 5 | 806 | 73 | |
| AK018758.2 | - | 4 - 5 | 847 | 73 | ||
| AK145674.1 | - | 4 - 5 | 778 | 73 | ||
| AK152899.1 | - | 4 - 5 | 1185 | 73 | ||
| AK154979.1 | - | 4 - 5 | 826 | 73 | ||
| AK170079.1 | - | 4 - 5 | 849 | 73 | ||
| AX799084.1 | - | 4 - 5 | 848 | 73 | ||
| BC052433.1 | - | 4 - 5 | 1137 | 6837097 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stt3b
Genomic Map