Assay Details
| Gene Symbol: Stt3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024222.2 | NP_077184.2 | 6 - 7 | 1336 | 86 | |
| GenBank mRNA | AK018758.2 | - | 6 - 7 | 1046 | 86 | |
| AK145674.1 | - | 6 - 7 | 977 | 86 | ||
| AK148065.1 | - | 6 - 7 | 1034 | 86 | ||
| AK152899.1 | - | 6 - 7 | 1384 | 86 | ||
| AK154979.1 | - | 6 - 7 | 1025 | 86 | ||
| AK170079.1 | - | 6 - 7 | 1048 | 86 | ||
| AX799084.1 | - | 6 - 7 | 1047 | 86 | ||
| BC003206.1 | - | 1 - 2 | 48 | 86 | ||
| BC052433.1 | - | 6 - 7 | 1336 | 6837097 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stt3b
Genomic Map