Assay Details
| Gene Symbol: Gadl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028638.1 | NP_082914.1 | 7 - 8 | 749 | 89 | |
| GenBank mRNA | BC052327.1 | - | 7 - 8 | 749 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gadl1
Genomic Map