Assay Details
| Gene Symbol: Gadl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028638.1 | NP_082914.1 | 10 - 11 | 992 | 62 | |
| GenBank mRNA | AK003937.1 | - | 1 - 2 | 36 | 62 | |
| BC052327.1 | - | 10 - 11 | 992 | 62 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gadl1
Genomic Map