Assay Details
Gene Symbol: Ambn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK017250.1 | - | 10 - 11 | 795 | 60 | |
BC087927.1 | - | 10 - 11 | 840 | 30504437 | 60 | |
U65021.1 | - | 10 - 11 | 775 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1999463 | U65021.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1999463 | AK017250.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1999463 | NM_009664.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 233270 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 765597 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 407626 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 506785 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 503917 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 507933 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 198847 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 37662 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P135359 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P151637 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1999463 | U65021.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1999463 | AK017250.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1999463 | NM_009664.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P340484 | U65021.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P340484 | AK017250.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P340484 | NM_009664.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P340484 | BC087927.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23872264 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23872273 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23872251 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23872252 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23872250 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23872265 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23872247 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23872255 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23872270 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23872253 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23872244 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23872259 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23872262 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23872248 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23872246 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23872268 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23872261 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23872266 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23872257 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23872260 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23872271 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P151637 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05646747 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01155063 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P135359 | - | - |
Whole Mouse Genome G4122A | A_51_P340484 | U65021.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P340484 | AK017250.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P340484 | NM_009664.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P340484 | BC087927.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139818 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113122 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P135359 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P151637 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04436313 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04436332 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04436305 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04436308 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04436319 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04436306 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872301 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872367 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872262 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872334 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872250 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872259 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872274 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872326 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872260 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872345 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872338 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872353 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872363 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872268 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872277 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872289 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872386 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872297 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872281 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872340 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872323 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872310 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872265 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872306 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872299 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872261 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872312 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872253 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872276 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872315 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872325 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872360 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872266 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872336 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872248 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872257 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872355 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872285 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872317 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872399 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872396 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872335 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872357 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872279 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872313 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872245 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872304 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872403 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872270 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872293 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872397 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872369 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872288 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872264 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872400 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872294 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872247 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872327 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872246 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872348 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872252 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872330 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872283 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872366 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872365 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872401 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872378 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872384 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872373 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872391 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872343 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872376 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872364 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872374 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872332 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872271 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872321 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23872255 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23872283 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23872297 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23872253 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23872248 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23872304 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23872380 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04436333 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23872377 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23872286 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04694832 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04436340 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04122574 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04219249 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23872294 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23872293 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04429468 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23872270 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23872262 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23872299 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04436305 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23872247 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23872268 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04122571 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23872277 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23872378 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23872264 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23872303 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23872271 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23872291 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04436318 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23872290 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23872279 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23872376 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23872281 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23872243 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23872389 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23872250 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23872261 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23872252 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23872266 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23872301 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04436317 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23872391 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04429470 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23872390 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23872257 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23872288 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23872255 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23872276 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23872285 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23872244 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04814144 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23872302 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10522959 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10522950 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10522958 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10522955 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10522948 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10522949 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10522954 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10522947 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10522960 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10522951 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10522953 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10522956 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10522957 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1449417_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10522946 | - | - |
Mouse Genome 430 2.0 Array | 1449417_at | - | - |
Mouse Genome 430 2.0 Array | 1449417_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11698_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11698_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0500007129.mm.1 | TC0500000811.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500007129.mm.1 | NM_009664.2 | Refseq |
Mouse Expression Set 430 | 1449417_PM_at | - | - |
Mouse Expression Set 430 | 1449417_at | - | - |
Murine Genome U74v2 Set | 93692_PM_f_at | - | - |
Murine Genome U74v2 Set | 93692_f_at | - | - |
Mouse Genome 430A 2.0 Array | 1449417_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1449417_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1449417_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1449417_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 4150170 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004150170 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1240933 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1240933 | - | - |
mouse-6 v1.1 (Illumina) | scl0011698.1_91-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4150170 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0011698.1_91-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4150170 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1240933 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1240933 | - | - |
