Assay Details
Gene Symbol: Ambn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK017250.1 | - | 1 - 2 | 108 | 65 | |
BC087927.1 | - | 1 - 2 | 108 | 30504437 | 65 | |
U65021.1 | - | 1 - 2 | 88 | 65 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC087927.1, U65021.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
