Assay Details
| Gene Symbol: Padi4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011061.2 | NP_035191.2 | 8 - 9 | 960 | 139 | |
| GenBank mRNA | AB013850.1 | - | 8 - 9 | 953 | 139 | |
| AK137625.1 | - | 8 - 9 | 962 | 139 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Padi4
Genomic Map