Assay Details
| Gene Symbol: Padi4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011061.2 | NP_035191.2 | 9 - 10 | 1065 | 67 | |
| GenBank mRNA | AB013850.1 | - | 9 - 10 | 1058 | 67 | |
| AK084904.1 | - | 3 - 4 | 2438 | 67 | ||
| AK137625.1 | - | 9 - 10 | 1067 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Padi4
Genomic Map