Assay Details
| Gene Symbol: Igsf21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198610.2 | NP_941012.1 | 3 - 4 | 743 | 75 | |
| GenBank mRNA | AK137439.1 | - | 3 - 4 | 737 | 75 | |
| BC055811.1 | - | 3 - 4 | 716 | 6414125 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Igsf21
Genomic Map