Assay Details
| Gene Symbol: Cdkl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276315.1 | NP_001263244.1 | 6 - 7 | 1347 | 74 | |
| NM_016912.2 | NP_058608.1 | 6 - 7 | 1081 | 74 | ||
| NM_177270.4 | NP_796244.2 | 6 - 7 | 1081 | 74 | ||
| GenBank mRNA | AB029065.1 | - | 6 - 7 | 1057 | 74 | |
| AB029066.1 | - | 6 - 7 | 1045 | 74 | ||
| AB029067.1 | - | 6 - 7 | 1335 | 74 | ||
| AK030598.1 | - | 6 - 7 | 1336 | 74 | ||
| AK139820.1 | - | 6 - 7 | 1289 | 74 | ||
| AK144574.1 | - | 6 - 7 | 1082 | 74 | ||
| AK145688.1 | - | 6 - 7 | 1230 | 74 | ||
| BC117894.1 | - | 6 - 7 | 990 | 40096852 | 74 | |
| BC117895.1 | - | 6 - 7 | 990 | 40096853 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdkl2
Genomic Map