Assay Details
| Gene Symbol: Naaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163687.1 | NP_001157159.1 | 3 - 4 | 554 | 76 | |
| NM_025972.4 | NP_080248.2 | 3 - 4 | 560 | 76 | ||
| GenBank mRNA | AB162194.1 | - | 3 - 4 | 526 | 76 | |
| AK008776.1 | - | 3 - 4 | 562 | 76 | ||
| AK017141.1 | - | 3 - 4 | 539 | 76 | ||
| AK133485.1 | - | 3 - 4 | 545 | 76 | ||
| AK133658.1 | - | 3 - 4 | 562 | 76 | ||
| AK146191.1 | - | 3 - 4 | 548 | 76 | ||
More Information
Set Membership: |
|

Naaa
Genomic Map