Assay Details
| Gene Symbol: Naaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163687.1 | NP_001157159.1 | 5 - 6 | 727 | 67 | |
| NM_025972.4 | NP_080248.2 | 5 - 6 | 733 | 67 | ||
| GenBank mRNA | AB162194.1 | - | 5 - 6 | 699 | 67 | |
| AK008776.1 | - | 5 - 6 | 735 | 67 | ||
| AK017141.1 | - | 5 - 6 | 712 | 67 | ||
| AK133485.1 | - | 5 - 6 | 718 | 67 | ||
| AK133658.1 | - | 5 - 6 | 735 | 67 | ||
| AK146191.1 | - | 5 - 6 | 721 | 67 | ||
| BC004572.1 | - | 5 - 6 | 703 | 3586255 | 67 | |
More Information
Set Membership: |
|

Naaa
Genomic Map