Assay Details
| Gene Symbol: Naaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163687.1 | NP_001157159.1 | 6 - 7 | 899 | 67 | |
| NM_025972.4 | NP_080248.2 | 6 - 7 | 905 | 67 | ||
| GenBank mRNA | AB162194.1 | - | 6 - 7 | 871 | 67 | |
| AK008776.1 | - | 6 - 7 | 907 | 67 | ||
| AK017141.1 | - | 6 - 7 | 882 | 67 | ||
| AK133485.1 | - | 6 - 7 | 890 | 67 | ||
| AK146191.1 | - | 6 - 7 | 893 | 67 | ||
| BC004572.1 | - | 6 - 7 | 875 | 3586255 | 67 | |
More Information
Set Membership: |
|

Naaa
Genomic Map