Assay Details
| Gene Symbol: Naaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163687.1 | NP_001157159.1 | 7 - 8 | 964 | 70 | |
| NM_025972.4 | NP_080248.2 | 7 - 8 | 970 | 70 | ||
| GenBank mRNA | AB162194.1 | - | 7 - 8 | 936 | 70 | |
| AK008776.1 | - | 7 - 8 | 972 | 70 | ||
| AK017141.1 | - | 7 - 8 | 947 | 70 | ||
| AK133485.1 | - | 7 - 8 | 955 | 70 | ||
| AK133658.1 | - | 7 - 8 | 971 | 70 | ||
| AK146191.1 | - | 7 - 8 | 958 | 70 | ||
| BC004572.1 | - | 7 - 8 | 940 | 3586255 | 70 | |
More Information
Set Membership: |
|

Naaa
Genomic Map