Assay Details
| Gene Symbol: Ror2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013846.3 | NP_038874.3 | 3 - 4 | 693 | 102 | |
| GenBank mRNA | AB010384.1 | - | 3 - 4 | 461 | 102 | |
| AK031112.1 | - | 3 - 4 | 663 | 102 | ||
| AK031789.1 | - | 3 - 4 | 451 | 102 | ||
| AK083538.1 | - | 3 - 4 | 669 | 102 | ||
| AK084752.1 | - | 3 - 4 | 659 | 102 | ||
| AK157705.1 | - | 3 - 4 | 709 | 102 | ||
| BC070436.1 | - | 3 - 4 | 693 | 30535615 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ror2
Genomic Map