Assay Details
| Gene Symbol: Ror2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013846.3 | NP_038874.3 | 4 - 5 | 726 | 117 | |
| GenBank mRNA | AB010384.1 | - | 4 - 5 | 494 | 117 | |
| AK031112.1 | - | 4 - 5 | 696 | 117 | ||
| AK031789.1 | - | 4 - 5 | 484 | 117 | ||
| AK083538.1 | - | 4 - 5 | 702 | 117 | ||
| AK084752.1 | - | 4 - 5 | 692 | 117 | ||
| AK157705.1 | - | 4 - 5 | 742 | 117 | ||
| BC070436.1 | - | 4 - 5 | 726 | 30535615 | 117 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ror2
Genomic Map