Assay Details
| Gene Symbol: Avl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030235.1 | NP_084511.1 | 14 - 15 | 1960 | 87 | |
| GenBank mRNA | AK020003.1 | - | 14 - 15 | 1937 | 87 | |
| AK083605.1 | - | 14 - 15 | 1917 | 87 | ||
| AK122228.1 | - | 14 - 15 | 1938 | 87 | ||
| AK147365.1 | - | 14 - 15 | 1962 | 87 | ||
| BC117878.1 | - | 14 - 15 | 1922 | 40095961 | 87 | |
| BC117879.1 | - | 14 - 15 | 1922 | 86 | ||
More Information
Set Membership: |
|

Avl9
Genomic Map