Assay Details
| Gene Symbol: Fgfr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008011.2 | NP_032037.2 | 10 - 11 | 1557 | 83 | |
| GenBank mRNA | AK084850.1 | - | 10 - 11 | 1557 | 83 | |
| AK085723.1 | - | 10 - 11 | 1555 | 83 | ||
| AY493377.2 | - | 9 - 10 | 1443 | 83 | ||
| BC033313.1 | - | 10 - 11 | 1744 | 5132831 | 83 | |
| DQ388428.1 | - | 9 - 10 | 1443 | 83 | ||
| X59927.1 | - | 10 - 11 | 1499 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fgfr4
Genomic Map